Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(2-Carboxyethyl)benzoic Acid Methyl Ester 95%

Supplier: AMBEED, INC
Warning

A771833-1G A771833-250MG A771833-5G A771833-25G
77159-026EA 42 USD InStock
77159-026 77159-028 77159-030 77686-786
4-(2-Carboxyethyl)benzoic Acid Methyl Ester 95%
4-(2-Carboxyethyl)benzoic Acid Methyl Ester
Precaución: for Research Use Only


Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD12406797
CAS Number: 151937-09-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty