Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-((1R,2R)-Cyclohexane-1,2-diyl)bis(4-methylbenzenesulfonamide) ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A773421-100G A773421-1G A773421-25G A773421-5G
77159-042EA 250.01 USD InStock
77159-042 77159-044 77159-046 77159-048
N,N'-((1R,2R)-Cyclohexane-1,2-diyl)bis(4-methylbenzenesulfonamide) ≥98%, ee 99%
N,N'-((1R,2R)-Cyclohexane-1,2-diyl)bis(4-methylbenzenesulfonamide)
Precaución: for Research Use Only


Formula: C₂₀H₂₆N₂O₄S₂
MDL Number: MFCD01321377
CAS Number: 143585-47-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty