Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Phenyl-5,7-dihydroindolo[2,3-b]carbazole 95%

Supplier: AMBEED, INC
Warning

A774973-1G A774973-5G
77159-058EA 29 USD InStock
77159-058 77159-060
5-Phenyl-5,7-dihydroindolo[2,3-b]carbazole 95%
5-Phenyl-5,7-dihydroindolo[2,3-b]carbazole
Precaución: for Research Use Only


Formula: C₂₄H₁₆N₂
MDL Number: MFCD29035249
CAS Number: 1448296-00-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty