Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trioctylphosphine oxide 98%

Supplier: AMBEED, INC
Danger

A811952-25G A811952-5G
77159-296EA 50 USD InStock
77159-296 77159-298
Trioctylphosphine oxide 98%
Trioctylphosphine oxide
Precaución: for Research Use Only


Formula: C₂₄H₅₁OP
MDL Number: MFCD00002083
CAS Number: 78-50-2
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty