Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(1-Imidazolyl)methyl]phenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A825468-1G A825468-250MG A825468-5G
77159-368EA 69 USD InStock
77159-368 77159-370 77159-372
4-[(1-Imidazolyl)methyl]phenylboronic acid pinacol ester 98%
4-[(1-Imidazolyl)methyl]phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₆H₂₁BN₂O₂
MW: 284.161 g/mol
MDL Number: MFCD18735902
CAS Number: 1351478-35-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty