Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium trifluoro(phenylethynyl)borate 98%

Supplier: AMBEED, INC
Warning

A854808-250MG A854808-5G
77159-524EA 26 USD InStock
77159-524 77159-526
Potassium trifluoro(phenylethynyl)borate 98%
Potassium trifluoro(phenylethynyl)borate
Precaución: for Research Use Only


Formula: C₈H₅BF₃·K
MDL Number: MFCD11052662
CAS Number: 485338-93-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty