Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 2-Fluorobenzylphosphonate 95%

Supplier: AMBEED, INC
Warning

A856831-100MG A856831-1G A856831-250MG A856831-5G
77159-528EA 20 USD InStock
77159-528 77159-530 77159-532 77159-534
Diethyl 2-Fluorobenzylphosphonate 95%
Diethyl 2-Fluorobenzylphosphonate
Precaución: for Research Use Only


Formula: C₁₁H₁₆FO₃P
MW: 246.215 g/mol
MDL Number: MFCD22574982
CAS Number: 63909-54-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty