Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Bromo-[1,1'-biphenyl]-4-carboxaldehyde 95%

Supplier: AMBEED, INC
Warning

A862034-100MG A862034-250MG A862034-1G
77159-550EA 12 USD InStock
77159-550 77159-552 77686-901
4'-Bromo-[1,1'-biphenyl]-4-carboxaldehyde 95%
4'-Bromo-[1,1'-biphenyl]-4-carboxaldehyde
Precaución: for Research Use Only


Formula: C₁₃H₉BrO
MW: 261.114 g/mol
MDL Number: MFCD05980925
CAS Number: 50670-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty