Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dibromoaniline 98%

Supplier: AMBEED, INC
Warning

A866854-25G A866854-5G A866854-1G
77159-568EA 69 USD InStock
77159-568 77159-570 77686-906
3,4-Dibromoaniline 98%
3,4-Dibromoaniline
Precaución: for Research Use Only


Formula: C₆H₅Br₂N
MDL Number: MFCD00041310
CAS Number: 615-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty