Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate 98%

Supplier: AMBEED, INC
Warning

A871876-10G A871876-1G A871876-250MG A871876-25G A871876-5G
77159-586EA 68 USD InStock
77159-586 77159-588 77159-590 77159-592 77159-594
tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate 98%
tert-Butyl 3,3-difluoro-4-oxopiperidine-1-carboxylate hydrate
Precaución: for Research Use Only


Formula: C₁₀H₁₅F₂NO₃·H₂O
MDL Number: MFCD19443979
CAS Number: 1400264-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty