Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Icilin 97%

Supplier: AMBEED, INC
Warning

A881912-10MG A881912-1MG A881912-25MG A881912-50MG
77159-648EA 46 USD InStock
77159-648 77159-650 77159-652 77159-654
Icilin 97%
Icilin
Precaución: for Research Use Only


Formula: C₁₆H₁₃N₃O₄
MW: 311.297 g/mol
MDL Number: MFCD01689072
CAS Number: 36945-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty