Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenhydramine hydrochloride 99%

Supplier: AMBEED, INC
Warning

A884689-100G A884689-250MG A884689-25G A884689-5G
77159-680EA 51 USD InStock
77159-680 77159-682 77159-684 77159-686
Diphenhydramine hydrochloride 99%
Diphenhydramine hydrochloride
Precaución: for Research Use Only


Formula: C₁₇H₂₁NO
MDL Number: MFCD00012479
CAS Number: 147-24-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty