Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TAK-242 98%

Supplier: AMBEED, INC
Warning

A888245-10MG A888245-1MG A888245-5MG A888245-50MG
77159-712EA 146 USD InStock
77159-712 77159-714 77159-716 77686-927
TAK-242 98%
TAK-242
Precaución: for Research Use Only


Formula: C₁₅H₁₇NO₄ClFS
MDL Number: MFCD11046362
CAS Number: 243984-11-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty