Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibenzyloxy benzaldehyde 96%

Supplier: AMBEED, INC
Warning

A893189-10G A893189-1G
77159-738EA 170 USD InStock
77159-738 77159-740
2,4-Dibenzyloxy benzaldehyde 96%
2,4-Dibenzyloxy benzaldehyde
Precaución: for Research Use Only


Formula: C₂₁H₁₈O₃
MDL Number: MFCD00022024
CAS Number: 13246-46-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty