Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-3-bromo-6-chloroindole 97%

Supplier: AMBEED, INC
Warning

A897576-10G A897576-1G A897576-250MG A897576-5G
77159-748EA 215 USD InStock
77159-748 77159-750 77159-752 77159-754
N-Boc-3-bromo-6-chloroindole 97%
N-Boc-3-bromo-6-chloroindole
Precaución: for Research Use Only


Formula: C₁₃H₁₃BrClNO₂
MDL Number: MFCD17169908
CAS Number: 1246471-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty