Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole-1,1-dioxide 95+%

Supplier: AMBEED, INC
Warning

A912660-100MG A912660-1G A912660-250MG A912660-5G
77159-842EA 13 USD InStock
77159-842 77159-844 77159-846 77159-848
(S)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole-1,1-dioxide 95+%
(S)-4,5,6-Tetrahydro-3H-pyrrolo[1,2-c]oxathiazole-1,1-dioxide
Precaución: for Research Use Only


Formula: C₅H₉NO₃S
MDL Number: MFCD22123262
CAS Number: 132635-95-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty