Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Monoelaidin 97+%

Supplier: AMBEED, INC
Warning

A918420-100MG A918420-1G A918420-250MG
77159-880EA 12 USD InStock
77159-880 77159-882 77159-884
Monoelaidin 97+%
Monoelaidin
Precaución: for Research Use Only


Formula: C₂₁H₄₀O₄
MDL Number: MFCD00069635
CAS Number: 2716-53-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty