Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((S)-Dec-1-yn-5-yloxy)tetrahydro-2H-pyran 95%

Supplier: AMBEED, INC
Warning

A948897-1G A948897-250MG A948897-5G
77159-976EA 64 USD InStock
77159-976 77159-978 77159-980
2-((S)-Dec-1-yn-5-yloxy)tetrahydro-2H-pyran 95%
2-((S)-Dec-1-yn-5-yloxy)tetrahydro-2H-pyran
Precaución: for Research Use Only


Formula: C₁₅H₂₆O₂
MDL Number: MFCD29037441
CAS Number: 223734-62-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty