Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

sec-Butylhydrazine dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A954343-100MG A954343-1G A954343-250MG
77160-006EA 20 USD InStock
77160-006 77160-008 77160-010
sec-Butylhydrazine dihydrochloride 95%
sec-Butylhydrazine dihydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₄Cl₂N₂
MDL Number: MFCD11840328
CAS Number: 1177361-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty