Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4,5,6-trichloroaniline 98%

Supplier: AMBEED, INC
Warning

A955572-10G A955572-1G A955572-25G
77160-020EA 41 USD InStock
77160-020 77160-022 77160-024
2-Bromo-4,5,6-trichloroaniline 98%
2-Bromo-4,5,6-trichloroaniline
Precaución: for Research Use Only


Formula: C₆H₃BrF₃N
MDL Number: MFCD00013388
CAS Number: 122375-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty