Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium pyrimidine-5-trifluoroborate 95+%

Supplier: AMBEED, INC
Warning

A962390-1G A962390-250MG A962390-5G
77160-026EA 31 USD InStock
77160-026 77160-028 77160-030
Potassium pyrimidine-5-trifluoroborate 95+%
Potassium pyrimidine-5-trifluoroborate
Precaución: for Research Use Only


Formula: C₄H₃BF₃KN₂
MDL Number: MFCD09992922
CAS Number: 1242733-91-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty