Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Carboethoxyphenyl cyclopropyl ketone 95%

Supplier: AMBEED, INC
Warning

A981814-100MG A981814-250MG
77160-092EA 97 USD InStock
77160-092 77160-094
4-Carboethoxyphenyl cyclopropyl ketone 95%
4-Carboethoxyphenyl cyclopropyl ketone
Precaución: for Research Use Only


Formula: C₁₃H₁₄O₃
MDL Number: MFCD03841215
CAS Number: 863769-67-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty