Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Chloroethyl)-1H-pyrazole 98%

Supplier: AMBEED, INC
Danger

A101506-1G A101506-250MG A101506-25G A101506-5G
77188-556EA 29 USD InStock
77188-556 77188-558 77188-560 77188-562
1-(2-Chloroethyl)-1H-pyrazole 98%
1-(2-Chloroethyl)-1H-pyrazole
Precaución: for Research Use Only


Formula: C₅H₇ClN₂
MDL Number: MFCD00173067
CAS Number: 96450-53-2
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty