Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-8-(trifluoromethyl)quinoline 97%

Supplier: AMBEED, INC
Warning

A105657-100MG A105657-1G
77160-466EA 52 USD InStock
77160-466 77160-468
5-Bromo-8-(trifluoromethyl)quinoline 97%
5-Bromo-8-(trifluoromethyl)quinoline
Precaución: for Research Use Only


Formula: C₁₀H₅BrF₃N
MDL Number: MFCD25542357
CAS Number: 1239460-75-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty