Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromophenoxy)-5-(trifluoromethyl)pyridine 97%

Supplier: AMBEED, INC
Warning

A105795-100MG A105795-10G A105795-1G A105795-250MG A105795-5G
77188-774EA 9 USD InStock
77188-774 77188-776 77188-778 77188-780 77188-782
2-(4-Bromophenoxy)-5-(trifluoromethyl)pyridine 97%
2-(4-Bromophenoxy)-5-(trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₁₂H₇BrF₃NO
MDL Number: MFCD00204096
CAS Number: 137736-63-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty