Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Bromomethyl)adamantane 98%

Supplier: AMBEED, INC
Warning

A108864-1G A108864-250MG A108864-25G A108864-5G
77188-952EA 13.39 USD InStock
77188-952 77188-954 77188-956 77188-958 77188-960
1-(Bromomethyl)adamantane 98%
1-(Bromomethyl)adamantane
Precaución: for Research Use Only


Formula: C₁₁H₁₇Br
MDL Number: MFCD00185303
CAS Number: 14651-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty