Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-cyanocyclopropanecarboxylate 97%

Supplier: AMBEED, INC
Warning

A112111-100G A112111-10G A112111-1G A112111-25G A112111-5G
77189-112EA 99 USD InStock
77189-112 77189-114 77189-116 77189-118 77189-120
Ethyl-1-cyanocyclopropanecarboxylate 97%
Ethyl-1-cyanocyclopropanecarboxylate
Precaución: for Research Use Only


Formula: C₇H₉NO₂
MW: 139.16 g/mol
MDL Number: MFCD00040159
CAS Number: 1558-81-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty