Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-1,2-Cyclohexanedicarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A112867-100G A112867-25G A112867-5G A112867-10G
77189-166EA 42 USD InStock
77189-166 77189-168 77189-170 77745-838
trans-1,2-Cyclohexanedicarboxylic acid 97%
trans-1,2-Cyclohexanedicarboxylic acid
Precaución: for Research Use Only


Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 339
MDL Number: MFCD00001464
CAS Number: 2305-32-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty