Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzocyclobutene-4-boronic acid 98%

Supplier: AMBEED, INC
Warning

A113063-100MG A113063-1G A113063-250MG A113063-5G
77160-850EA 15 USD InStock
77160-850 77160-852 77160-854 77160-856
Benzocyclobutene-4-boronic acid 98%
Benzocyclobutene-4-boronic acid
Precaución: for Research Use Only


Formula: C₈H₇BO₂
MW: 145.951 g/mol
MDL Number: MFCD09752664
CAS Number: 195730-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty