Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,6-Dichlorohexane 95%

Supplier: AMBEED, INC
Danger

A113188-25G A113188-100G A113188-500G
77189-176EA 5 USD InStock
77189-176 77745-839 77745-840
1,6-Dichlorohexane 95%
1,6-Dichlorohexane
Precaución: for Research Use Only


Formula: ClH₂C(CH₂)₄CH₂Cl
MDL Number: MFCD00001020
CAS Number: 2163-00-0
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty