Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Torin 1 98%

Supplier: AMBEED, INC
Warning

A113194-10MG A113194-5MG
77189-178EA 112 USD InStock
77189-178 77189-180
Torin 1 98%
Torin 1
Precaución: for Research Use Only


Formula: C₃₅H₂₈F₃N₅O₂
MDL Number: MFCD18782653
CAS Number: 1222998-36-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty