Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,2-Dimethoxy-N-methylacetamide 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A113296-10G A113296-1G A113296-250MG A113296-25G A113296-5G A113296-100G
77160-866EA 50 USD InStock
77160-866 77160-868 77160-870 77160-872 77160-874 77683-033
N,2-Dimethoxy-N-methylacetamide 95%, Ambeed.Inc
N,2-Dimethoxy-N-methylacetamide
Precaución: For research use only.


Formula: C₅H₁₁NO₃
MW: 133.146 g/mol
MDL Number: MFCD11848891
CAS Number: 132289-57-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty