Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Clevudine 98%

Supplier: AMBEED, INC
Warning

A113960-10MG A113960-1MG A113960-5MG
77189-212EA 26 USD InStock
77189-212 77189-214 77189-216
Clevudine 98%
Clevudine
Precaución: for Research Use Only


Formula: C₁₀H₁₃FN₂O₅
MDL Number: MFCD00935785
CAS Number: 163252-36-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty