Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-methylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A114293-100G A114293-10G A114293-1G A114293-25G A114293-5G
77189-226EA 119 USD InStock
77189-226 77189-228 77189-230 77189-232 77189-234
4-Fluoro-3-methylphenylboronic acid 98%
4-Fluoro-3-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₈BFO₂
MDL Number: MFCD01863527
CAS Number: 139911-27-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty