Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiopicolinamide 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A114642-100G A114642-10G A114642-25G A114642-5G A114642-1G
77160-902EA 77 USD InStock
77160-902 77160-904 77160-906 77160-908 77683-044
Thiopicolinamide 97%, Ambeed.Inc
Thiopicolinamide
Precaución: For research use only.


Formula: C₆H₆N₂S
MDL Number: MFCD00087576
CAS Number: 5346-38-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty