Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Bis(trimethylsilyl)benzene 97%

Supplier: AMBEED, INC
Warning

A115083-100G A115083-10G A115083-1G A115083-250MG A115083-25G A115083-5G
77189-318EA 185 USD InStock
77189-318 77189-320 77189-322 77189-324 77189-326 77189-328
1,4-Bis(trimethylsilyl)benzene 97%
1,4-Bis(trimethylsilyl)benzene
Precaución: for Research Use Only


Formula: C₁₂H₂₂Si₂
MW: 222.48
Boiling Pt: 83
Melting Pt: 97.5
MDL Number: MFCD00015592
CAS Number: 13183-70-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty