Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

MK-3102 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A115550-100MG A115550-10MG A115550-50MG A115550-5MG A115550-250MG
77160-976EA 66 USD InStock
77160-976 77160-978 77160-980 77160-982 77160-984 77160-986 77683-050
MK-3102 98%, Ambeed.Inc
MK-3102
Precaución: For research use only.


Formula: C₁₇H₂₀F₂N₄O₃S
MW: 398.43 g/mol
MDL Number: MFCD22573261
CAS Number: 1226781-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty