Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene 98%

Supplier: AMBEED, INC
Warning

A116105-10G A116105-1G A116105-5G
77189-360EA 42 USD InStock
77189-360 77189-362 77189-364 77189-366
1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene 98%
1-[(4-Methoxyphenyl)ethynyl]-4-n-propylbenzene
Precaución: for Research Use Only


Formula: C₁₈H₁₈O
MDL Number: MFCD04038795
CAS Number: 39969-26-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty