Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methylpyridazine-3-carboximidamide acetate 97%

Supplier: AMBEED, INC
Warning

A117374-1G A117374-250MG A117374-5G
77189-456EA 96 USD InStock
77189-456 77189-458 77189-460
6-Methylpyridazine-3-carboximidamide acetate 97%
6-Methylpyridazine-3-carboximidamide acetate
Precaución: for Research Use Only


Formula: C₈H₁₂N₄O₂
MDL Number: MFCD28024743
CAS Number: 1609406-73-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty