Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminobenzothiazol-6-ol 97%

Supplier: AMBEED, INC
Warning

A117407-100G A117407-1G A117407-250MG A117407-25G A117407-5G
77161-072EA 240 USD InStock
77161-072 77161-074 77161-076 77161-078 77161-080
2-Aminobenzothiazol-6-ol 97%
2-Aminobenzothiazol-6-ol
Precaución: for Research Use Only


Formula: C₇H₆N₂OS
MDL Number: MFCD00511763
CAS Number: 26278-79-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty