Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(2,2,2-Trifluoroethoxy)picolinic acid 97%

Supplier: AMBEED, INC
Warning

A117651-100MG A117651-10G A117651-1G
77189-470EA 56 USD InStock
77189-470 77189-472 77189-474
6-(2,2,2-Trifluoroethoxy)picolinic acid 97%
6-(2,2,2-Trifluoroethoxy)picolinic acid
Precaución: for Research Use Only


Formula: C₈H₆F₃NO₃
MW: 221.13 g/mol
MDL Number: MFCD16697752
CAS Number: 1247503-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty