Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α,6-Tetrafluoro-m-toluic acid 98%

Supplier: AMBEED, INC
Warning

A118828-100G A118828-10G A118828-1G A118828-25G A118828-5G
77189-512EA 74 USD InStock
77189-512 77189-514 77189-516 77189-518 77189-520
α,α,α,6-Tetrafluoro-m-toluic acid 98%
α,α,α,6-Tetrafluoro-m-toluic acid
Precaución: for Research Use Only


Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
MDL Number: MFCD00061292
CAS Number: 115029-23-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty