Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Bis(dimethylsilyl)benzene 97%

Supplier: AMBEED, INC
Warning

A118888-100G A118888-10G A118888-5G
77189-530EA 223 USD InStock
77189-530 77189-532 77189-534
1,4-Bis(dimethylsilyl)benzene 97%
1,4-Bis(dimethylsilyl)benzene
Precaución: for Research Use Only


Formula: C₁₀H₁₈Si₂
MW: 194.42
MDL Number: MFCD00039790
CAS Number: 2488-01-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty