Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine 98%

Supplier: AMBEED, INC
Warning

A119381-100G A119381-1G A119381-25G
77189-576EA 1045 USD InStock
77189-576 77189-578 77189-580
(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine 98%
(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
Precaución: for Research Use Only


Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
MDL Number: MFCD06858462
CAS Number: 122536-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty