Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethylphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A119449-10G A119449-25G A119449-5G
77189-582EA 18 USD InStock
77189-582 77189-584 77189-586
2,5-Dimethylphenylboronic acid 97%
2,5-Dimethylphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₁₁BO₂
MW: 149.98
MDL Number: MFCD01863525
CAS Number: 85199-06-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty