Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dacomitinib 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A119719-1MG A119719-5G
77161-216EA 8.24 USD InStock
77161-216 77161-218 77161-220 77683-075
Dacomitinib 98%, Ambeed.Inc
Dacomitinib
Precaución: For research use only.


Formula: C₂₄H₂₅ClFN₅O₂
MDL Number: MFCD19443734
CAS Number: 1110813-31-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty