Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl p-Tolyl sulfone 98%

Supplier: AMBEED, INC
Danger

A120919-1000G A120919-100G A120919-10G A120919-25G A120919-500G
77189-662EA 59 USD InStock
77189-662 77189-664 77189-666 77189-668 77189-670
Methyl p-Tolyl sulfone 98%
Methyl p-Tolyl sulfone
Precaución: for Research Use Only


Formula: C₈H₁₀O₂S
MDL Number: MFCD00014742
CAS Number: 3185-99-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty