Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Formyl-4,5-dimethoxybenzonitrile 98%

Supplier: AMBEED, INC
Warning

A121196-1G A121196-250MG
77161-312EA 142 USD InStock
77161-312 77161-314
2-Formyl-4,5-dimethoxybenzonitrile 98%
2-Formyl-4,5-dimethoxybenzonitrile
Precaución: for Research Use Only


Formula: C₁₀H₉NO₃
MDL Number: MFCD24678322
CAS Number: 1013112-48-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty