Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4,6-Dichloro-2-(ethoxycarbonyl)-1H-indol-3-yl)acrylic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A122014-100MG A122014-1G
77161-346EA 110 USD InStock
77161-346 77683-097
3-(4,6-Dichloro-2-(ethoxycarbonyl)-1H-indol-3-yl)acrylic acid 95%, Ambeed.Inc
3-(4,6-Dichloro-2-(ethoxycarbonyl)-1H-indol-3-yl)acrylic acid
Precaución: For research use only.


Formula: C₁₄H₁₁Cl₂NO₄
MDL Number: MFCD04114763
CAS Number: 159054-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty