Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium 1-cyano-3-ethoxy-2,3-dioxopropan-1-ide 95%

Supplier: AMBEED, INC
Warning

A122850-1G A122850-250MG A122850-5G
77161-428EA 25 USD InStock
77161-428 77161-430 77161-432
Potassium 1-cyano-3-ethoxy-2,3-dioxopropan-1-ide 95%
Potassium 1-cyano-3-ethoxy-2,3-dioxopropan-1-ide
Precaución: for Research Use Only


Formula: C₆H₆KNO₃
MDL Number: MFCD28016957
CAS Number: 92664-05-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty